missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rubidium chloride, 99% (metals basis), Alfa Aesar™

Testing for perchloric acid, source of rubidium, catalyst

Manufacturer:  Alfa Aesar 1289222

 View more versions of this product

Catalog No. AA1289222



Specifications

Specifications

Rubidium chloride
99% (metals basis)
ClRb
14,8284
Soluble in water; Practically insoluble in alcohol
[Cl-].[Rb+]
120.92
CHEBI:78672
Crystalline
2.800g/mL
1.493
1390°C
718°C
7791-11-9
(metals basis)
MFCD00011187
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
rubidium(1+);chloride
62683
120.92
99%
Odorless
Hygroscopic
White
100g

Chemical Identifiers

7791-11-9
120.92
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
62683
rubidium(1+);chloride
ClRb
MFCD00011187
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
CHEBI:78672
[Cl-].[Rb+]
Safety and Handling

Safety and Handling

RTECSNumber : VL8575000

TSCA : TSCA

Documents