missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Rubidium chloride, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 201270250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1390.0°C | |
718.0°C | |
White | |
25 g | |
ClRb | |
MFCD00011187 | |
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride | |
FGDZQCVHDSGLHJ-UHFFFAOYSA-M | |
rubidium(1+);chloride | |
62683 | |
120.92 | |
Glass bottle |
Rubidium chloride | |
7791-11-9 | |
Crystalline Powder and/or Crystals | |
99+% | |
RbCl | |
15, 8417 | |
Solubility in water: 910g/L (20 c). Other solubilities: 1g/0.7mL boiling water, practically insoluble in alcohols | |
[Cl-].[Rb+] | |
120.92 | |
CHEBI:78672 | |
≥99% | |
2.8g/mL |
Chemical Identifiers
7791-11-9 | |
120.92 | |
FGDZQCVHDSGLHJ-UHFFFAOYSA-M | |
62683 | |
[Cl-].[Rb+] |
ClRb | |
MFCD00011187 | |
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride | |
CHEBI:78672 |
Safety and Handling
EINECSNumber : 232-240-9
RUO – Research Use Only