missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rubidium chloride, 99.975% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 1085109

 View more versions of this product

Catalog No. AA1085109



Specifications

Specifications

Rubidium chloride
99.975% (metals basis)
MFCD00011187
rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rubidium-chlorid, rubidium-chloride, rubidiumchloride, unii-n3shc5273s
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
rubidium(1+);chloride
62683
120.92
2.8 g/cm3 at 20°C
1.493
1390°C
718°C
7791-11-9
ClRb
White crystals
Soluble in water.
[Cl-].[Rb+]
120.92
CHEBI:78672
Crystalline Aggregates
Odorless
Hygroscopic
White
10g

Chemical Identifiers

7791-11-9
120.92
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
62683
rubidium(1+);chloride
ClRb
MFCD00011187
rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rubidium-chlorid, rubidium-chloride, rubidiumchloride, unii-n3shc5273s
CHEBI:78672
[Cl-].[Rb+]
Safety and Handling

Safety and Handling

EINECSNumber : 232-240-9

RTECSNumber : VL8575000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents