Please login to your online account to display your discounted pricing

Alfa Aesar™ Rubidium chloride, 99.8% (metals basis)

$87.74 - $87.74

Chemical Identifiers

Chemical Name or Material Rubidium chloride
CAS 7791-11-9
Molecular Formula Rb
Linear Formula RbCl
Formula Weight 120.92
MDL Number MFCD00011187
Merck Index 14,8284
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1056318

  SDS
alfa aesar™
1056318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1056309

  SDS
alfa aesar
1056309
10g Each for $87.74
Description & Specifications

Specifications

Formula Weight 120.92
Assay Percent Range 99.8% (metals basis)
Molecular Formula Rb
Chemical Name or Material Rubidium chloride
TSCA TSCA
Mesh Size −20 Mesh
Odor Odorless
Physical Form Crystalline
Refractive Index 1.493
Assay 99.8%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
CAS 7791-11-9
Merck Index 14,8284
Boiling Point 1390°C
Melting Point 718°C
Quantity 50g
Density 2.800g/mL
RTECS Number VL8575000
Linear Formula RbCl
MDL Number MFCD00011187
Color White