Please login to your online account to display your discounted pricing

Rubidium carbonate, 99%, pure, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC317151000

 View more versions of this product

Catalog No. AC317151000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5% (ex Rb)
Physical Form Powder
Formula Weight 230.95
Quantity 100g
Color White
Purity Grade Pure
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Rubidium carbonate
Molecular Formula CO3Rb2
Linear Formula Rb2CO3
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
MDL Number MFCD00011188
Packaging Glass bottle
Health Hazard 1 Warning
Assay Percent Range 99%
Name Note 99%
CAS 584-09-8

Specifications

Chemical Name or Material Rubidium carbonate
Name Note 99%
CAS 584-09-8
Purity Grade Pure
Assay ≥98.5% (ex Rb)
Assay Percent Range 99%
Molecular Formula CO3Rb2
Linear Formula Rb2CO3
Formula Weight 230.95
MDL Number MFCD00011188
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents