Please login to your online account to display your discounted pricing

Rubidium carbonate, 99%, pure, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  317150250

 View more versions of this product

Catalog No. AC317150250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5% (ex Rb)
Physical Form Powder
Formula Weight 230.95
Quantity 25g
Color White
Purity Grade Pure
Chemical Name or Material Rubidium carbonate
Name Note 99%
Packaging Glass bottle
Assay Percent Range 99%
Molecular Formula CO3Rb2
Linear Formula Rb2CO3
MDL Number MFCD00011188
CAS 584-09-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Rubidium carbonate
Name Note 99%
CAS 584-09-8
Purity Grade Pure
Assay ≥98.5% (ex Rb)
Assay Percent Range 99%
Molecular Formula CO3Rb2
Linear Formula Rb2CO3
Formula Weight 230.95
MDL Number MFCD00011188
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents