missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rubidium carbonate, 99.975% (metals basis), Alfa Aesar™ Puratronic™

Manufacturer:  Thermo Fisher Scientific 1110503

 View more versions of this product

Catalog No. AA1110503



Specifications

Specifications

Rubidium carbonate
99.975% (metals basis)
MFCD00011188
carbonic acid dirubidium salt
C(=O)([O-])[O-].[Rb+].[Rb+]
230.944
230.95
Odorless
1g
584-09-8
CO3Rb2
White powder
WPFGFHJALYCVMO-UHFFFAOYSA-L
rubidium(1+);carbonate
11431
Powder
White

Chemical Identifiers

584-09-8
230.944
WPFGFHJALYCVMO-UHFFFAOYSA-L
11431
C(=O)([O-])[O-].[Rb+].[Rb+]
CO3Rb2
MFCD00011188
carbonic acid dirubidium salt
rubidium(1+);carbonate
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.

H315-H319-H335-H303: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May be harmful if swallowed.

EINECSNumber : 209-530-9

Documents