missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rubidium carbonate, 99.975% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 1110514

 View more versions of this product

Catalog No. AA1110514



Description

Description

Specifications

Specifications

Rubidium carbonate
99.975% (metals basis)
MFCD00011188
carbonic acid dirubidium salt, carbonic acid, dirubidium salt, dirubidium 1+ ion carbonate, dirubidium carbonate, dirubidium monocarbonate, potassium carbonate anhydrous, rubidium 1+ carbonate, rubidium carbonate, rubidiumcarbonate
WPFGFHJALYCVMO-UHFFFAOYSA-L
rubidium(1+);carbonate
11431
Powder
Very Hygroscopic
837°C
584-09-8
CO3Rb2
White powder
Soluble in water at 20°C 4500g/L.
C(=O)([O-])[O-].[Rb+].[Rb+]
230.944
230.95
Odorless
White
25g

Chemical Identifiers

584-09-8
230.944
WPFGFHJALYCVMO-UHFFFAOYSA-L
11431
C(=O)([O-])[O-].[Rb+].[Rb+]
CO3Rb2
MFCD00011188
carbonic acid dirubidium salt, carbonic acid, dirubidium salt, dirubidium 1+ ion carbonate, dirubidium carbonate, dirubidium monocarbonate, potassium carbonate anhydrous, rubidium 1+ carbonate, rubidium carbonate, rubidiumcarbonate
rubidium(1+);carbonate
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 209-530-9

RTECSNumber : FG0650000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents