missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rubidium carbonate, 99%

Manufacturer:  Alfa Aesar™ A1224006

 View more versions of this product

Catalog No. AAA1224006


 

Description

Description

Applications
Rubidium carbonate is used as a raw materials for preparation of rubidium metal and various rubidium salts, for the manufacturing of special glass, for the manufacturing of high energy density micro cells and crystal scintillation counters. It is also used as a part of a catalyst for preparing short-chain alcohols from feed gas.

Solubility
Soluble in water4500 g/L @20°C.

Notes
Hygroscopic. Store under dry inert gas. Store away from bases, water/ moisture.
Specifications

Specifications

Rubidium carbonate
99%
Rb2CO3
WPFGFHJALYCVMO-UHFFFAOYSA-L
rubidium(1+);carbonate
11431
Odorless
5g
584-09-8
CO3Rb2
MFCD00011188
C(=O)([O-])[O-].[Rb+].[Rb+]
230.944
230.95
837°C

Chemical Identifiers

584-09-8
230.944
WPFGFHJALYCVMO-UHFFFAOYSA-L
CO3Rb2
MFCD00011188
11431
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : FG0650000

TSCA : TSCA

SDS Documents