Please login to your online account to display your discounted pricing

Alfa Aesar™ Rubidium bromide, 99.8% (metals basis)

Chemical Identifiers

Chemical Name or Material Rubidium bromide
CAS 7789-39-1
Formula Weight 165.38
MDL Number MFCD00011186
Merck Index 14,8283
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1289106

  SDS
alfa aesar™
1289106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1289114

  SDS
alfa aesar™
1289114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1289122

  SDS
alfa aesar™
1289122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 165.38
Chemical Name or Material Rubidium bromide
TSCA TSCA
Odor Odorless
Physical Form Crystalline
Refractive Index 1.5530
Assay 99.8%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
MDL Number MFCD00011186
CAS 7789-39-1
Merck Index 14,8283
Boiling Point 1340°C
Melting Point 693°C
Quantity 5g
Solubility Information Soluble in water