missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rubidium acetate, 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 1289009

 View more versions of this product

Catalog No. AA1289009



Specifications

Specifications

Rubidium acetate
246°C
563-67-7
C2H3O2Rb
rubidium acetate, acetic acid, rubidium salt, unii-86h795sz6d, acetic acid, rubidium salt 1:1, acetic acid rubidium, rubidio acetate, acmc-1aks3, rubidium 1+ acetate ion, rubidium 1+ ion acetate, c2h3o2.rb
CC(=O)[O-].[Rb+]
144.512
144.52
99.8%
Hygroscopic
10g
(metals basis)
MFCD00013058
FOGKDYADEBOSPL-UHFFFAOYSA-M
rubidium(1+);acetate
23673628
Crystalline

Chemical Identifiers

563-67-7
144.512
FOGKDYADEBOSPL-UHFFFAOYSA-M
23673628
CC(=O)[O-].[Rb+]
C2H3O2Rb
MFCD00013058
rubidium acetate, acetic acid, rubidium salt, unii-86h795sz6d, acetic acid, rubidium salt 1:1, acetic acid rubidium, rubidio acetate, acmc-1aks3, rubidium 1+ acetate ion, rubidium 1+ ion acetate, c2h3o2.rb
rubidium(1+);acetate
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents