missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rubidium, 99.75% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1031503

 View more versions of this product

Catalog No. AA1031503



Specifications

Specifications

Rubidium
7440-17-7
1.532g/cm3
Rb
atom
[Rb]
Odorless
5357696
Solid
1g
Silver-colored solid
99.75% (Metals basis)
MFCD00134055
IGLNJRXAVVLDKE-UHFFFAOYSA-N
rubidium
85.468
CHEBI:33322
Silver

Chemical Identifiers

7440-17-7
85.468
IGLNJRXAVVLDKE-UHFFFAOYSA-N
5357696
rubidium
Rb
MFCD00134055
atom
CHEBI:33322
[Rb]
Safety and Handling

Safety and Handling

H260-H314-H318: In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 4.3, Packaging Group: I

EINECSNumber : 231-126-6

Documents