missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rubidium, 99.75% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1031522

 View more versions of this product

Catalog No. AA1031522

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Rubidium
7440-17-7
99.75% (Metals basis)
MFCD00134055
IGLNJRXAVVLDKE-UHFFFAOYSA-N
rubidium
85.468
CHEBI:33322
Silver
100g
Silver-colored solid
Rb
atom, mlt4718tjw, rubidio, rubidium, isotope of mass 103, rubidium, isotope of mass 106, rubidium-103, rubidium-106, un1423 dangerous when wet, unii-mlt4718tjw
[Rb]
Odorless
5357696
Vacuum sealed in breakseal ampoules

Chemical Identifiers

7440-17-7
85.468
IGLNJRXAVVLDKE-UHFFFAOYSA-N
5357696
rubidium
Rb
MFCD00134055
atom, mlt4718tjw, rubidio, rubidium, isotope of mass 103, rubidium, isotope of mass 106, rubidium-103, rubidium-106, un1423 dangerous when wet, unii-mlt4718tjw
CHEBI:33322
[Rb]
Safety and Handling

Safety and Handling

P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c

H260-H314-H335

DOTInformation : DOT Class: 4.3, Packaging Group: I

EINECSNumber : 231-126-6

RTECSNumber : VL8500000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents