Please login to your online account to display your discounted pricing

Rubicene, 98%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: fisher scientific acros  333782500

 View more versions of this product

Catalog No. AC333782500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 326.4
Quantity 250mg
Color Red
Beilstein 05, III, 2673
EINECS Number 205-899-5
Chemical Name or Material Rubicene
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C26H14
MDL Number MFCD00021273
CAS 197-61-5
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Rubicene
Name Note 98%
CAS 197-61-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C26H14
Formula Weight 326.4
MDL Number MFCD00021273