Please login to your online account to display your discounted pricing

Rubicene, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Rubicene
Name Note 98%
CAS 197-61-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C26H14
Formula Weight 326.4
MDL Number MFCD00021273
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC333782500

  SDS
fisher scientific acros
333782500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC333781000

  SDS
thermo fisher scientific
333781000
Glass bottle 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 326.4
Physical Form Powder
Quantity 250mg
Color Red
Beilstein 05, III, 2673
EINECS Number 205-899-5
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C26H14
CAS 197-61-5
MDL Number MFCD00021273
Name Note 98%
Assay Percent Range 98%
Chemical Name or Material Rubicene