Rubbermaid™ Cleanroom Maintenance Mops
Looped-end, tailbanded mop for greater floor coverage with each stroke
Manufacturer: Rubbermaid™ T30000WH00
Description
- For use in ISO Class 7 (Class 10,000) and higher industrial applications such as paint booths, pharmaceutical labs, etc.
- Made of a woven, rayon/polyester fabric that will not fray or lint
- Launderable and pre-shrunk for long product life
- 5 in. (12.7cm) universal headband
Specifications
White | |
35 to 41 in. | |
Polyester, Rayon |
Mop handles, class 10,000 and higher applications | |
5.5 to 6.5 in. | |
Cleanroom Mop |