missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rubbermaid™ Cleanroom Maintenance Mops

Looped-end, tailbanded mop for greater floor coverage with each stroke

$73.36 - $94.24

Specifications

Color White
Material Polyester, Rayon
Type Cleanroom Mop
For Use With (Application) Mop handles, class 10,000 and higher applications
Width (English) 5.5 to 6.5 in.
View More Specs

Products 2
Catalog Number Mfr. No. Length (English) Price Quantity  
Catalog Number Mfr. No. Length (English) Price Quantity  
19-076-533
View Documents
Rubbermaid™
RCPT30100WH00
41 to 47 in.
Each for $94.24
Case of 12 Each for $1,061.28
 
19-076-532
View Documents
Rubbermaid™
T30000WH00
35 to 41 in.
Each for $73.36
Case of 12 Each for $826.20
 
Description

Description

  • For use in ISO Class 7 (Class 10,000) and higher industrial applications such as paint booths, pharmaceutical labs, etc.
  • Made of a woven, rayon/polyester fabric that will not fray or lint
  • Launderable and pre-shrunk for long product life
  • 5 in. (12.7cm) universal headband

Specifications

Specifications

White
Cleanroom Mop
5.5 to 6.5 in.
Polyester, Rayon
Mop handles, class 10,000 and higher applications
SDS