GSC Go Science Crazy Rubberized Cloth Apron
Provides basic protection from chemicals in laboratory or classroom settings. The GSC International Rubberized Cloth Apron is made of rubberized cloth and available in several sizes.
Manufacturer:  GSC Go Science Crazy 12760
 View more versions of this product
Catalog No. S13707
Alert:
Intended for ages 13 and up. May contain latex.
Description
Description
  • Great for laboratory and classroom safety
  • Material is rubberized and resistant to most common chemicals
  • Cloth tie strings provide steadiness and flexibility
  • Available in several sizes
Specifications
Specifications
Laboratory or classroom settings
27 x 36 in.
Rubberized Cloth
Cloth tie strings
Documents