missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Walter Products Rubberized Chemical Resistant Aprons
Flexible and lightweight.
Manufacturer: Walter Products AB2430
Description
- Made of chemically resistant rubberized material
- Reinforced at points of strain
- Ties at waist and neck
- Latex-free
- Color: black
Specifications
Rubberized apron | |
Black | |
30 in. | |
24 in. |
Rubberized Cloth | |
Yes | |
76.2 cm | |
60.96 cm |