Please login to your online account to display your discounted pricing
Rubber Stoppers for Kjeldahl Flasks
Light tan rubber stoppers
$77.09 - $362.42
Products
Catalog Number Mfr. No. Price Quantity    
14-149B
the plasticoid company
500M181
Pack of 18 Each for $77.09
Case of 5 Pack for $385.45
14-149A
the plasticoid company
500M180
Case of 5 Each for $362.42
Description & Specifications
  • Choice of styles: solid or with one hole of proper size for Pyrex™ Kjeldahl Connecting Bulbs
  • One-hole style can be enlarged or the solid stopper bored to take larger sizes
  • Top diameter: 32mm; bottom diameter: 26mm; height: 34mm; one-hole bore diameter, 8mm
†The approximate per-pack quantity shown is based on the average number of stoppers in 1 lb. (454g).