United Scientific Supplies Rubber Stopper Assortment
Assorted styles, Size No. 1 to 6
Manufacturer:  United Scientific Supplies RSTPK1
Catalog No. S67820
Description
Description
  • These black stoppers are made of solid natural rubber that stays pliable over a long period of time.
  • Ideal for use with non-aggressive or concentrated solutions.
  • Low sulfur content. Withstand temperatures from –25 to 70°C (–13 to 158°F).
  • Each stopper has its size marked in raised figures.
  • All stoppers are 25 mm tall.
  • Sold as one-pound bags.
  • Contains an assortment of 1-hole stoppers, Size No. 00 - 7
Specifications
Specifications
Rubber
Non-sterile
Various
Various
Laboratory use
Autoclavable
Various
Various
Assorted Style Stopper
Assorted
Various
Various
1 to 6
Black
Various