Eppendorf™ Rubber Cushion Tube Adapters for Eppendorf™ Fixed-Angle Rotors
For use with Eppendorf 5702 Fixed-Angle Rotors
Manufacturer: Eppendorf™ 022639455
Description
- For use with fixed-angle rotors 05-400-304 and 05-400-305
- Sold in packs of 10
Specifications
Rubber | |
Eppendorf 5702 Centrifuges with fixed-angle rotor F-35-30-17 | |
15mL round-bottom tubes | |
Cushion adapter |