missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ru(OAc)2[(S)-tolbinap], TCI America™
$68.02 - $222.30
Chemical Identifiers
CAS | 106681-15-6 |
---|---|
Molecular Formula | C52H48O4P2Ru |
Molecular Weight (g/mol) | 899.97 |
MDL Number | MFCD09753021 |
InChI Key | OXESSJMVRGFBNX-UHFFFAOYSA-N |
Synonym | Diacetato[(S)-(-)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl]ruthenium(II) |
PubChem CID | 11007467 |
IUPAC Name | bis(acetic acid) {2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane ruthenium |
SMILES | [Ru].CC(O)=O.CC(O)=O.CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
R0169200MG
|
TCI America
R0169200MG |
200 mg |
Each for $68.02
|
|
|||||
R01691G
|
TCI America
R01691G |
1 g |
Each for $222.30
|
|
|||||
Chemical Identifiers
106681-15-6 | |
899.97 | |
OXESSJMVRGFBNX-UHFFFAOYSA-N | |
11007467 | |
[Ru].CC(O)=O.CC(O)=O.CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1 |
C52H48O4P2Ru | |
MFCD09753021 | |
Diacetato[(S)-(-)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl]ruthenium(II) | |
bis(acetic acid) {2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane ruthenium |
Specifications
106681-15-6 | |
C52H48O4P2Ru | |
200 mg | |
OXESSJMVRGFBNX-UHFFFAOYSA-N | |
bis(acetic acid) {2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane ruthenium | |
11007467 | |
Crystalline Powder |
Black-Yellow | |
MFCD09753021 | |
Diacetato[(S)-(-)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl]ruthenium(II) | |
[Ru].CC(O)=O.CC(O)=O.CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1 | |
899.97 | |
897.95 | |
Ru(OAc)2[(S)-tolbinap] |
Safety and Handling
TSCA : No