missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ru(OAc)2[(R)-binap], TCI America™

Manufacturer:  Tci America R01661G

 View more versions of this product

Catalog No. R01661G



Specifications

Specifications

Ru(OAc)2[(R)-binap]
1g
C48H40O4P2Ru
Diacetato[(R)-(+)-2,2′C-bis(diphenylphosphino)-1,1′C-binaphthyl]ruthenium(II)
CC(=O)O.CC(=O)O.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.[Ru]
843.862
841.85
Brown-Yellow
325146-81-4
MFCD09753020
XANQJWCSQJDBEA-UHFFFAOYSA-N
acetic acid;[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;ruthenium
118705203
Crystal-Powder at 20°C

Chemical Identifiers

325146-81-4
843.862
XANQJWCSQJDBEA-UHFFFAOYSA-N
118705203
CC(=O)O.CC(=O)O.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.[Ru]
C48H40O4P2Ru
MFCD09753020
Diacetato[(R)-(+)-2,2′C-bis(diphenylphosphino)-1,1′C-binaphthyl]ruthenium(II)
acetic acid;[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;ruthenium
Safety and Handling

Safety and Handling

TSCA : No

Documents