Please login to your online account to display your discounted pricing

(RS)-MCPG, Tocris

Non-selective mGlu antagonist

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material (RS)-MCPG
Assay Percent Range >99%
Molecular Formula C10H11NO4
Formula Weight 209.2
Synonym (RS)-α-Methyl-4-carboxyphenylglycine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

033610

 
r & d systems
RDS033610
10mg Each for $177.37

033650

 
r & d systems
RDS033650
50mg Each for $746.48
Description & Specifications

Specifications

Chemical Name or Material (RS)-MCPG
CAS 146669-29-6
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C10H11NO4
Formula Weight 209.2
Synonym (RS)-α-Methyl-4-carboxyphenylglycine
Quantity 10mg
Safety and Handling

Store at Room Temperature