Please login to your online account to display your discounted pricing

(RS)-CPP, Tocris

Potent NMDA antagonist

$223.30 - $891.94

Chemical Identifiers

Chemical Name or Material (RS)-CPP
Assay Percent Range >98%
Molecular Formula C8H17N2O5P
Formula Weight 252.21
Synonym (RS)-3-(2-Carboxypiperazin-4-yl)-propyl-1-phosphonic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

017310

 
r & d systems
017310
10mg Each for $223.30

017350

 
r & d systems
017350
50mg Each for $891.94
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 252.21
Assay Percent Range >98%
Recommended Storage Desiccate at Room Temperature
Synonym (RS)-3-(2-Carboxypiperazin-4-yl)-propyl-1-phosphonic acid
Molecular Formula C8H17N2O5P
CAS 100828-16-8
Chemical Name or Material (RS)-CPP
Safety and Handling

Desiccate at Room Temperature