Please login to your online account to display your discounted pricing

(RS)-APICA, Tocris


Selective group II antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS107310

 View more versions of this product

Catalog No. 107310

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 257.18
Quantity 10mg
Molecular Formula C10H12NO5P
Assay Percent Range >98%
CAS 170847-18-4
Chemical Name or Material (RS)-APICA
Synonym (RS)-1-Amino-5-phosphonoindan-1-carboxylic acid
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material (RS)-APICA
Assay Percent Range >98%
Molecular Formula C10H12NO5P
Formula Weight 257.18
Synonym (RS)-1-Amino-5-phosphonoindan-1-carboxylic acid
Accessories

Desiccate at +4°C