Please login to your online account to display your discounted pricing

(RS)-APICA, Tocris

Selective group II antagonist

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material (RS)-APICA
Assay Percent Range >98%
Molecular Formula C10H12NO5P
Formula Weight 257.18
Synonym (RS)-1-Amino-5-phosphonoindan-1-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

107310

 
r & d systems
RDS107310
10mg Each for $188.60

107350

 
r & d systems
RDS107350
50mg Each for $794.96
Description & Specifications

Specifications

Formula Weight 257.18
Quantity 10mg
Molecular Formula C10H12NO5P
Assay Percent Range >98%
CAS 170847-18-4
Chemical Name or Material (RS)-APICA
Synonym (RS)-1-Amino-5-phosphonoindan-1-carboxylic acid
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C