Please login to your online account to display your discounted pricing

RS 79948 hydrochloride, Tocris


Potent, selective α2 antagonist

Quantity 50mg

Manufacturer: r & d systems  RDS098750

 View more versions of this product

Catalog No. 098750

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 400.96
Chemical Name or Material RS 79948 hydrochloride
Assay Percent Range >99%
Molecular Formula C19H28N2O3S·HCl
CAS 186002-54-0
Synonym (8aR,12aS,13aS)-5,8,8a,9,10,11,12,12a,13,13a-dechydro-3-methoxy-12-(ethylsulfonyl)-6H-isoquino[2,1-g][1,6]naphthyridine hydrochloride
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material RS 79948 hydrochloride
Assay Percent Range >99%
Molecular Formula C19H28N2O3S·HCl
Formula Weight 400.96
Synonym (8aR,12aS,13aS)-5,8,8a,9,10,11,12,12a,13,13a-dechydro-3-methoxy-12-(ethylsulfonyl)-6H-isoquino[2,1-g][1,6]naphthyridine hydrochloride
Accessories

Store at Room Temperature