Please login to your online account to display your discounted pricing

RS 67506 hydrochloride, Tocris


5-HT4 partial agonist

Quantity 10mg

Manufacturer: r & d systems  RDS099010

 View more versions of this product

Catalog No. 099010

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 454.41
Assay Percent Range >98%
CAS 168986-61-6
Molecular Formula C18H28ClN3O4S·HCl
Synonym 1-(4-Amino-5-chloro-2-methoxyphenyl)-3-[1-2-methylsulphonylamino)ethyl-4-piperidinyl]-1-propanone hydrochloride
Chemical Name or Material RS 67506 hydrochloride
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material RS 67506 hydrochloride
Assay Percent Range >98%
Molecular Formula C18H28ClN3O4S·HCl
Formula Weight 454.41
Synonym 1-(4-Amino-5-chloro-2-methoxyphenyl)-3-[1-2-methylsulphonylamino)ethyl-4-piperidinyl]-1-propanone hydrochloride
Accessories

Store at Room Temperature