Please login to your online account to display your discounted pricing

RS 67506 hydrochloride, Tocris

5-HT4 partial agonist

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material RS 67506 hydrochloride
Assay Percent Range >98%
Molecular Formula C18H28ClN3O4S·HCl
Formula Weight 454.41
Synonym 1-(4-Amino-5-chloro-2-methoxyphenyl)-3-[1-2-methylsulphonylamino)ethyl-4-piperidinyl]-1-propanone hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

099050

 
r & d systems
RDS099050
50mg Each for $682.67

099010

 
r & d systems
RDS099010
10mg Each for $164.61
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 454.41
Synonym 1-(4-Amino-5-chloro-2-methoxyphenyl)-3-[1-2-methylsulphonylamino)ethyl-4-piperidinyl]-1-propanone hydrochloride
Chemical Name or Material RS 67506 hydrochloride
Assay Percent Range >98%
CAS 168986-61-6
Molecular Formula C18H28ClN3O4S·HCl
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature