Please login to your online account to display your discounted pricing

RS 67333 hydrochloride, Tocris

5-HT4 partial agonist

$179.63 - $750.85

Chemical Identifiers

Chemical Name or Material RS 67333 hydrochloride
Assay Percent Range >99%
Molecular Formula C19H29ClN2O2·HCl
Formula Weight 389.36
Synonym 1-(4-Amino-5-chloro-2-methoxyphenyl)-3-[1-butyl-4-piperidinyl]-1-propanone hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

098950

 
r & d systems
RDS098950
50mg Each for $750.85

098910

 
r & d systems
RDS098910
10mg Each for $179.63
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 389.36
CAS 168986-60-5
Recommended Storage Store at Room Temperature
Chemical Name or Material RS 67333 hydrochloride
Assay Percent Range >99%
Molecular Formula C19H29ClN2O2·HCl
Synonym 1-(4-Amino-5-chloro-2-methoxyphenyl)-3-[1-butyl-4-piperidinyl]-1-propanone hydrochloride
Safety and Handling

Store at Room Temperature