Please login to your online account to display your discounted pricing

RS 56812 hydrochloride, Tocris


5-HT3 partial agonist

Quantity 50mg

Manufacturer: r & d systems  098850

 View more versions of this product

Catalog No. 098850

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material RS 56812 hydrochloride
CAS 143339-12-2
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C18H21N3O2·HCl
Formula Weight 347.84
Synonym (R)-N-(1-Azabicyclo[2.2.2]oct-3-yl)-2-(1-methyl-1H-indol-3-yl)-2-(1-methyl-1H-indol-3-yl)-2-oxoacetamide hydrochloride
Quantity 50mg

Specifications

Chemical Name or Material RS 56812 hydrochloride
Assay Percent Range >99%
Molecular Formula C18H21N3O2·HCl
Formula Weight 347.84
Synonym (R)-N-(1-Azabicyclo[2.2.2]oct-3-yl)-2-(1-methyl-1H-indol-3-yl)-2-(1-methyl-1H-indol-3-yl)-2-oxoacetamide hydrochloride
Accessories

Store at Room Temperature