Please login to your online account to display your discounted pricing

RS 56812 hydrochloride, Tocris

5-HT3 partial agonist

$172.26 - $726.06

Chemical Identifiers

Chemical Name or Material RS 56812 hydrochloride
Assay Percent Range >99%
Molecular Formula C18H21N3O2·HCl
Formula Weight 347.84
Synonym (R)-N-(1-Azabicyclo[2.2.2]oct-3-yl)-2-(1-methyl-1H-indol-3-yl)-2-(1-methyl-1H-indol-3-yl)-2-oxoacetamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

098810

 
r & d systems
098810
10mg Each for $172.26

098850

 
r & d systems
098850
50mg Each for $726.06
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 347.84
Chemical Name or Material RS 56812 hydrochloride
Assay Percent Range >99%
CAS 143339-12-2
Synonym (R)-N-(1-Azabicyclo[2.2.2]oct-3-yl)-2-(1-methyl-1H-indol-3-yl)-2-(1-methyl-1H-indol-3-yl)-2-oxoacetamide hydrochloride
Molecular Formula C18H21N3O2·HCl
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature