Please login to your online account to display your discounted pricing

RS 45041-190 hydrochloride, Tocris

High affinity I2 ligand; Highly selective

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material RS 45041-190 hydrochloride
Assay Percent Range >98%
Molecular Formula C11H12ClN3·HCl
Formula Weight 258.15
Synonym 4-Chloro-2-(imidazolin-2-yl)isoindoline hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

088910

 
r & d systems
RDS088910
10mg Each for $214.12

088950

 
r & d systems
RDS088950
50mg Each for $895.76
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 258.15
Assay Percent Range >98%
Chemical Name or Material RS 45041-190 hydrochloride
CAS 204274-74-8
Synonym 4-Chloro-2-(imidazolin-2-yl)isoindoline hydrochloride
Molecular Formula C11H12ClN3·HCl
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature