missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ (RS)-4-Carboxyphenylglycine

Broad spectrum EAA ligand

Manufacturer:  Tocris™ RDS032150

 View more versions of this product

Catalog No. 032150


 

Disclaimers:

For Research Use Only


Specifications

Description

Inhibitor, Enzyme; Tocris; (RS)-4-Carboxyphenylglycine; Broad spectrum EAA ligand; Molecular wt: 195.17; CAS Number: 7292-81-1; Formula: C9H9NO4; Purity:>99; 50mg

Broad spectrum EAA ligand. Separate isomers (R)-4-Carboxyphenylglycine and (S)-4-Carboxyphenylglycine also available (Cat. Nos. 0322 and 0323 respectively).
  • Formula: C9H9NO4
  • CAS Number: 7292-81-1
  • Solubility: Soluble to 100mM in 1eq. NaOH and to 5mM in water
  • Purity: >99
Description & Specifications

Specifications

(RS)-4-Carboxyphenylglycine
>99
50mg
(RS)-4-Carboxyphenylglycine
195.17