Please login to your online account to display your discounted pricing

Tocris™ (RS)-4-Carboxyphenylglycine

Broad spectrum EAA ligand

$175.98 - $741.73

Specifications

Product Type (RS)-4-Carboxyphenylglycine
Inhibitors (RS)-4-Carboxyphenylglycine
Molecular Weight 195.17
Purity >99
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

032110

 
tocris™
RDS032110
10mg Each for $175.98

032150

 
tocris™
RDS032150
50mg Each for $741.73
Description & Specifications

Specifications

Product Type (RS)-4-Carboxyphenylglycine
Inhibitors (RS)-4-Carboxyphenylglycine
Molecular Weight 195.17
Purity >99

Broad spectrum EAA ligand. Separate isomers (R)-4-Carboxyphenylglycine and (S)-4-Carboxyphenylglycine also available (Cat. Nos. 0322 and 0323 respectively).

  • Formula: C9H9NO4
  • CAS Number: 7292-81-1
  • Solubility: Soluble to 100mM in 1eq. NaOH and to 5mM in water
  • Purity: >99