Please login to your online account to display your discounted pricing

(RS)-3-Hydroxyphenylglycine, Tocris

Group I mGlu agonist

$175.98 - $741.73

Chemical Identifiers

Chemical Name or Material (RS)-3-Hydroxyphenylglycine
Assay Percent Range >98%
Molecular Formula C8H9NO3
Formula Weight 167.16
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

032410

 
r & d systems
RDS032410
10mg Each for $175.98

032450

 
r & d systems
RDS032450
50mg Each for $741.73
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 167.16
Chemical Name or Material (RS)-3-Hydroxyphenylglycine
Assay Percent Range >98%
Molecular Formula C8H9NO3
CAS 31932-87-3
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature