Please login to your online account to display your discounted pricing

RS 25344 hydrochloride, Tocris

Potent PDE4 inhibitor

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material RS 25344 hydrochloride
Assay Percent Range >98%
Molecular Formula C19H13N5O4·HCl
Formula Weight 411.8
Synonym 1-(3-Nitrophenyl)-3-(4-pyridinylmethyl)-pyrido[2,3-d]pyrimidine-2,3-(1H,3H)-dione hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

331510

 
r & d systems
331510
10mg Each for $201.36

331550

 
r & d systems
331550
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 411.8
Quantity 10mg
Molecular Formula C19H13N5O4·HCl
CAS 152815-28-6
Synonym 1-(3-Nitrophenyl)-3-(4-pyridinylmethyl)-pyrido[2,3-d]pyrimidine-2,3-(1H,3H)-dione hydrochloride
Chemical Name or Material RS 25344 hydrochloride
Assay Percent Range >98%
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature