Please login to your online account to display your discounted pricing

RS 23597-190 hydrochloride, Tocris

5-HT4 antagonist

$168.15 - $697.40

Chemical Identifiers

Chemical Name or Material RS 23597-190 hydrochloride
Assay Percent Range >98%
Molecular Formula C16H23ClN2O3·HCl
Formula Weight 363.28
Synonym 3-(Piperidin-1-yl)propyl 4-amino-5-chloro-2-methoxybenzoate hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

072810

 
r & d systems
RDS072810
10mg Each for $168.15

072850

 
r & d systems
RDS072850
50mg Each for $697.40
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 363.28
CAS 149719-06-2
Chemical Name or Material RS 23597-190 hydrochloride
Assay Percent Range >98%
Molecular Formula C16H23ClN2O3·HCl
Synonym 3-(Piperidin-1-yl)propyl 4-amino-5-chloro-2-methoxybenzoate hydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature