Please login to your online account to display your discounted pricing

RS 17053 hydrochloride, Tocris


α1A antagonist

Quantity 50mg

Manufacturer: r & d systems  098550

 View more versions of this product

Catalog No. 098550

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 449.42
CAS 169505-93-5
Synonym (N-[2-(2-Cyclopropylmethoxyphenoxy)ethyl]-5-chloro-α, α-dimethyl-1H-indole-3-ethanamine) hydrochloride
Chemical Name or Material RS 17053 hydrochloride
Molecular Formula C24H29N2O2Cl·HCl
Assay Percent Range >98%
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material RS 17053 hydrochloride
Assay Percent Range >98%
Molecular Formula C24H29N2O2Cl·HCl
Formula Weight 449.42
Synonym (N-[2-(2-Cyclopropylmethoxyphenoxy)ethyl]-5-chloro-α, α-dimethyl-1H-indole-3-ethanamine) hydrochloride
Accessories

Store at Room Temperature