Please login to your online account to display your discounted pricing

RS 16566 dihydrochloride, Tocris

5-HT3 ligand; Also shows affinity for zacopride binding site

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material RS 16566 dihydrochloride
Assay Percent Range >98%
Molecular Formula C18H23ClN4O·2HCl
Formula Weight 419.78
Synonym (R)-N-1-Azabicyclo[2.2.2]oct-3-yl-6-chloro-1-(1-methylethyl)-1H-benzimidazole-4-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

101550

 
r & d systems
101550
50mg Each for $904.70

101510

 
r & d systems
101510
10mg Each for $221.01
Description & Specifications

Specifications

Formula Weight 419.78
Quantity 50mg
Chemical Name or Material RS 16566 dihydrochloride
Molecular Formula C18H23ClN4O·2HCl
CAS 1217788-97-0
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Synonym (R)-N-1-Azabicyclo[2.2.2]oct-3-yl-6-chloro-1-(1-methylethyl)-1H-benzimidazole-4-carboxamide
Safety and Handling

Store at Room Temperature