Please login to your online account to display your discounted pricing

RS 127445 hydrochloride, Tocris

Selective, high affinity 5-HT2B antagonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material RS 127445 hydrochloride
Assay Percent Range >98%
Molecular Formula C17H16FN3·HCl
Formula Weight 317.79
Synonym 4-(4-Fluoro-1-naphthalenyl)-6-(1-methylethyl)-2-pyrimidinamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

299310

 
r & d systems
299310
10mg Each for $239.63

299350

 
r & d systems
299350
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 317.79
Quantity 10mg
Chemical Name or Material RS 127445 hydrochloride
CAS 199864-86-3
Synonym 4-(4-Fluoro-1-naphthalenyl)-6-(1-methylethyl)-2-pyrimidinamine hydrochloride
Assay Percent Range >98%
Molecular Formula C17H16FN3·HCl
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C