Please login to your online account to display your discounted pricing

RS 102221 hydrochloride, Tocris


Selective 5-HT2C antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS105010

 View more versions of this product

Catalog No. 105010

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 649.08
Quantity 10mg
Assay Percent Range >98%
Molecular Formula C27H31F3N4O7S·HCl
CAS 187397-18-8
Chemical Name or Material RS 102221 hydrochloride
Synonym 8-[5-(2,4-Dimethoxy-5-(4-trifluoromethylphenylsulphonamido)phenyl-5-oxopentyl]-1,3,8-triazaspiro[4.5]decane-2,4-dione hydrochloride
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material RS 102221 hydrochloride
Assay Percent Range >98%
Molecular Formula C27H31F3N4O7S·HCl
Formula Weight 649.08
Synonym 8-[5-(2,4-Dimethoxy-5-(4-trifluoromethylphenylsulphonamido)phenyl-5-oxopentyl]-1,3,8-triazaspiro[4.5]decane-2,4-dione hydrochloride
Accessories

Store at Room Temperature