Please login to your online account to display your discounted pricing

RS 100329 hydrochloride, Tocris

Potent, subtype-selective α1A antagonist

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material RS 100329 hydrochloride
Assay Percent Range >99%
Molecular Formula C20H25F3N4O3·HCl
Formula Weight 462.89
Synonym 5-Methyl-3-[3-[3-[4-[2-(2,2,2,-trifluroethoxy)phenyl]-1-piperazinyl]propyl]-2,4-(1H,3H)-pyrimidinedione hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

132510

 
r & d systems
132510
10mg Each for $221.01

132550

 
r & d systems
132550
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 462.89
Quantity 10mg
Chemical Name or Material RS 100329 hydrochloride
Molecular Formula C20H25F3N4O3·HCl
CAS 1215654-26-4
Synonym 5-Methyl-3-[3-[3-[4-[2-(2,2,2,-trifluroethoxy)phenyl]-1-piperazinyl]propyl]-2,4-(1H,3H)-pyrimidinedione hydrochloride
Assay Percent Range >99%
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature