Invitrogen™ RPS6KB2 (p70S6K Beta) Recombinant Human Protein DFS Item
RPS6KB2 (p70S6K beta) is a serine/threonine kinase with 2 non-identical kinase catalytic domains.
Manufacturer:  Invitrogen™ LSPV4055
 View more versions of this product
Catalog No. PV4055
Description
Description
RPS6KB2 (p70S6K beta) phosphorylates the S6 ribosomal protein and eukaryotic translation initiation factor 4B (eIF4B). Phosphorylation of S6 leads to an increase in protein synthesis and cell proliferation.
Specifications
Specifications
NP_003943
Frozen
82.2kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
RSK (RPS6K) Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
RPS6KB2 (p70S6K Beta)
Baculovirus
Human
GST Tag
Documents