missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ RPS6KB1 (p70S6K) Recombinant Human Protein
RPS6KB1 (p70S6K) is a serine/threonine kinase with 2 non-identical kinase catalytic domains.
$2650.00 - $11260.00
Specifications
Accession Number | NP_003152,T412E |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 75.1kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PR7310B
|
Invitrogen™
PR7310B |
100μg |
Each for $2,650.00
|
|
|||||
PV4054
|
Invitrogen™
PV4054 |
1mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
RPS6KB1 (p70S6K) phosphorylates several residues of the S6 ribosomal protein.
Specifications
NP_003152,T412E | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
RSK (RPS6K) Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
75.1kDa | |
RUO - research use only | |
RPS6KB1 (p70S6K) | |
Human | |
GST Tag | |
Frozen |