Invitrogen™ RPS6KB1 (p70S6K) Recombinant Human Protein DFS Item
RPS6KB1 (p70S6K) is a serine/threonine kinase with 2 non-identical kinase catalytic domains.
Manufacturer:  Invitrogen™ LSPV4054
 View more versions of this product
Catalog No. PV4054
Description
Description
RPS6KB1 (p70S6K) phosphorylates several residues of the S6 ribosomal protein.
Specifications
Specifications
NP_003152,T412E
Frozen
75.1kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Catalytic
Kinase Assay
RSK (RPS6K) Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
RPS6KB1 (p70S6K)
Baculovirus
Human
GST Tag
Documents