Invitrogen™ RPS6KA6 (RSK4) Recombinant Human Protein DFS Item
RPS6KA6 (RSK4) is a member of the RSK kinases family.
Manufacturer:  Invitrogen™ LSPV4558
 View more versions of this product
Catalog No. PV4558
Description
Description
RSK family members are activated by ERKs and mitogens and are unusual in that they contain 2 non-identical kinase domains.
Specifications
Specifications
NP_055311
Frozen
111.3kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
RSK (RPS6K) Kinase Family
Yes
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
RPS6KA6 (RSK4)
Baculovirus
Human
GST Tag
Documents