Invitrogen™ RPS6KA4 (MSK2) Recombinant Human Protein DFS Item
RPS6KA4 (MSK2) is a serine/threonine protein kinase that is essential for the regulation of various cellular processes.
Manufacturer:  Invitrogen™ LSPV4052
 View more versions of this product
Catalog No. PV4052
Description
Description
The RPS6KA4 (MSK2) kinase contains 2 non-identical kinase catalytic domains and phosphorylates various substrates, including CREB1 and c-fos.
Specifications
Specifications
NP_003933
Frozen
113.1kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
RSK (RPS6K) Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
RPS6KA4 (MSK2)
Baculovirus
Human
GST Tag
Documents